IFN gamma ELISA Kit, Human, Thermo Scientific
Supplier: Thermo Scientific
Assay IFN-γ (human interferon gamma) levels in human serum, plasma (EDTA, heparin, sodium citrate), and culture supernatant samples with our Thermo Scientific Human IFN gamma ELISA Kits.
Features of the Human IFN-γ ELISA Kit: †
Target: natural and recombinant human interferon gamma (IFNγ)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: absorbance at 450 nm minus absorbance at 550 nm
Assay Range: 25.6 to 1000 pg/mL
Sensitivity: ≤ 2 pg/mL
Precision: intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sample Types: cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3.5 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with with human IL-1α, IL-1β, IL-2, IL-3, IL-6, IL-7, IL-8, TNFα or GM-CSF
These human IFN-gamma ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interferon gamma (IFNγ) in samples at 50 to 100 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kit also contains a precoated 96-well strip plate. All components undergo quality control testing to minimize lot-to-lot variability.
Target: natural and recombinant human interferon gamma (IFNγ)
Ordering information: For Research Use Only. Not for use in diagnostic procedures. Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8˚C. See product manual for detailed contents and storage conditions for maximum stability.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...