Sodium Deoxycholate Detergent, Thermo Scientific
Supplier: Thermo Scientific
89904 89905
CAPI89904EA 639.99 CAD
CAPI89904 CAPI89905
Sodium Deoxycholate Detergent, Thermo Scientific
Cleaning Agents Detergents and Surface Cleansers
Deoxycholate is an ionic detergent that is especially useful for disrupting and dissociating protein interactions. Stored in a cool, dry, and well-ventilated area, protected from direct sunlight.
  • Popular anionic, bile-acid detergent for many laboratory uses
  • Effective in disrupting and dissociating many types of protein interaction
  • Can be removed from solution by dialysis
  • Useful for elution or regeneration of certain kinds of affinity columns
  • High-purity compound with low UV absorptivity
Sodium deoxycholate (deoxycholic acid) is a water-soluble, bile-acid, ionic detergent commonly used in protein methods
The effectiveness of a detergent in any application depend upon the detergent's concentration. Too much or too little detergent can often have a deleterious effect. It is recommended that you examine a variety of detergent concentrations in your application. At concentrations above 2 mM, cholate will form micelles having MW
2000. The small micelle size allows easy removal by dialysis or gel filtration when needed. Note: removal of a detergent from a protein solution may result in protein precipitation and/or aggregation.
Sodium deoxycholate is the detergent recommended for stripping endotoxin (Lipopolysaccharide or LPS) from immobilized Polymyxin B columns. This is the recommended product for use with the Thermo Scientific Detoxi-Gel Endotoxin Removing Gel.
Caution: For Research Use Only. Not for use in diagnostic procedures.