Pierce™ ActivX™ Serine Hydrolase Probes
Supplier: Thermo Scientific
88316 88317 88318
CAPI88316EA 1581.36 CAD
CAPI88316 CAPI88317 CAPI88318
Pierce™ ActivX™ Serine Hydrolase Probes
Assays Enzyme Assays
Thermo Scientific Serine Hydrolase Probes are ActivX™ Fluorophosphonate (FP) and other tagged phosphonate probes to purify or assay serine hydrolase active sites using fluorescence, biotin-affinity, or mass spectrometry.
  • Specifically Labels the Reactive Site of Active Serine Hydrolases
  • Compatible Tags Available for Capture, Detection and Staudinger Conjugation
  • Use for in vitro or Intracellular Enzyme Labeling
Azido-FP probes are used in combination with phosphine- or alkyne-derivatised tags for detection or enrichment.
Desthiobiotin-FP probes are used for enrichment and detection of active site-labeled proteins by Western blot or mass spectrometry.
TAMRA-FP probes are used to label and detect serine hydrolase activity in samples using fluorescent gel imaging, capillary electrophoresis or mass spectrometry.
Caution: For Research Use Only. Not for use in diagnostic procedures.