Imject™ Pierce™ Maleimide-Activated Carrier Proteins, Thermo Scientific
Supplier: Thermo Scientific
77605 77606 77610
CAPI77605EA 3624.08 CAD
CAPI77605 CAPI77606 CAPI77610
Imject™ Pierce™ Maleimide-Activated Carrier Proteins, Thermo Scientific
Protein Extraction Protein Extraction Kits
Imject® Maleimide-activated mcKLH and PEGylated mcKLH enable conjugation of sulfhydryl-containing peptide haptens to elicit an immune response and antibody production against the hapten
  • Used to achieve the highest antigen conjugation efficiency
  • Cysteine reactive chemistry enables controlled orientation of your antigen
  • High solubility minimises loss during conjugation
The mcKLH is maleimide-activated using a heterobifunctional crosslinker that contains an N-hydroxysuccinimide (NHS) ester and a maleimide group. The PEGylated mcKLH is activated with NHS-PEG2-Maleimide, creating a more soluble maleimideactivated mcKLH. Maleimide groups react with free sulfhydryls, forming stable thioether bonds.
Ordering information: Upon receipt store product at −20 °C. Product is shipped with an ice pack.
Caution: For Research Use Only. Not for use in diagnostic procedures.