Pierce™ Fast Western Blot Kit, ECL Substrate, Thermo Scientific
Supplier: Thermo Scientific
Pierce
35055 35055 35050 35050
CAPI35050EA 645.89 CAD
CAPI35050 CAPI35055 PI35050 PI35055
Pierce™ Fast Western Blot Kit, ECL Substrate, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
The Fast Western Blot Kit, ECL Substrate contains optimized reagents that shorten the time which is required to perform a typical chemiluminescent Western blot from 4 hours down to approximately 55 minutes.
  • Fast - all the sensitivity of Pierce ECL substrate and saving 4 to 5 hours per blot
  • No expensive hardware or vacuum required; no clogging issues
  • Optimized protocol makes Western blot analysis easier
  • No expensive consumables or extra equipment required
  • Kit stable for 1 year stored at 4°C
This complete kit provides all the reagents for blocking, antibody dilution and to complete a Western blot being probed with a mouse or rabbit primary antibody. The protocol requires minimal hands on time and yields results comparable to classic Western blotting with ECL. The included ECL substrate produces a chemiluminescent signal, which is detected using photographic or other imaging methods. Blots can be repeatedly exposed to film to obtain optimal results or stripped of the primary antibody and immunodetection reagents and reprobed.
Caution: For Research Use Only. Not for use in diagnostic procedures.