SuperSignal™ West Pico Complete Mouse IgG Detection Kit, Thermo Scientific
Supplier: Thermo Scientific
Thermo Scientific SuperSignal West Pico Complete IgG Detection Kit is a a kit containing goat anti-mouse or anti-rabbit secondary antibody HRP conjugate for the detection of mouse or rabbit primary antibody
- Optimized - secondary antibodies are validated for use with the SuperSignal West Pico Chemiluminescent Substrate, eliminating the need to test multiple dilutions
- Large packages - kits provide sufficient reagents to complete up to 50 experiments with standard mini gel blots (4000 square centimeters of membrane)
- Standard or complete - choose a kit with substrate and antibody only, or a complete kit that also includes TBS wash buffer and high-performance SuperBlock Blocking Buffer
All the components of these Western blotting kits are optimized for compatibility the SuperSignal West Pico Chemiluminescent Substrate. The kits are offered in a standard format (substrate and optimized HRP-conjugates) and a complete format that also includes the Thermo Scientific SuperBlock Buffer and BupH Tris Buffered Saline.
The kit is ideal for busy researchers or new beginners to Western blot analysis.
Caution: For Research Use Only. Not for use in diagnostic procedures.