Ammonium persulfate (APS), Pierce™

Supplier: Thermo Scientific

Synonyms: Ammonium persulfatePeroxidisulphuric acid diammonium saltAmmonium persulphatePeroxidisulfuric acid diammonium salt

17874
CAPI17874PK 272.82 CAD
CAPI17874
Ammonium persulfate (APS), Pierce™
Ammonium peroxodisulphate (APS)

Thermo Scientific Pierce Ammonium Persulfate (APS) is an oxidizing agent that is used with TEMED to catalyze the polymerization of acrylamide and bisacrylamide to prepare polyacrylamide gels for electrophoresis.


Purity: ≥98.0%


Insolubles: ≤0.005%


Ordering information: For Research Use Only. Not for use in diagnostic procedures.

Formula: (NH₄)₂S₂O₈
MW: 228.2 g/mol
MDL Number: MFCD00003390
CAS Number: 7727-54-0
UN: 1444
ADR: 5.1,III
https://ca.vwr.com/stibo/web/std.lang.all/06/39/16640639.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR