Pierce™ Agarose I (Molecular Biology Grade), Thermo Scientific
Supplier: Thermo Scientific
17850 17852
CAPI17850EA 816.03 CAD
CAPI17850 CAPI17852
Pierce™ Agarose I (Molecular Biology Grade), Thermo Scientific
Electrophoresis Gels Agarose and Acrylamide
Thermo Scientific Pierce Agaroses are highly purified and carefully blended formulations of regular that provide uniform lot-to-lot consistency for preparation of electrophoresis gels.
  • Standard melting temperature agarose for electrophoresis
  • Form: Powdered
Caution: For Research Use Only. Not for use in diagnostic procedures.