Pierce™ Renilla Luciferase Glow Assay Kit, Thermo Scientific
Supplier: Thermo Scientific
16166 16167
CAPI16166EA 410.02 CAD
CAPI16166 CAPI16167
Pierce™ Renilla Luciferase Glow Assay Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Pierce Renilla Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life approx
  • Sensitive—highly sensitive detection of Green Renilla luciferase activity
  • Stable—increased signal stability compared to flash assays
  • Compatible—assay reagents compatible with other Renilla luciferases
  • Automation-friendly—amenable to high throughput assays
  • Convenient—contains a universal cell lysis buffer and optimized glow assay reagent
3 hours), especially in the presence of Green Renilla luciferase reporter.
Delivery information: Kit contains Renilla Glow Assay Buffer (5 mL); 100X Coelenterazine (50 µL); and 2X Cell Lysis Buffer (6 mL).
Caution: For Research Use Only. Not for use in diagnostic procedures.