Pierce™ Gaussia Luciferase Glow Assay Kit, Thermo Scientific

Supplier: Thermo Scientific

16160 16161
CAPI16160EA 398.33 CAD
CAPI16160 CAPI16161
Pierce™ Gaussia Luciferase Glow Assay Kit, Thermo Scientific
Assays Enzyme Assays

The Thermo Scientific Pierce Gaussia luciferase glow assay kit provides an extremely bright and stable bioluminescence signal (half-life greater than 1 hour), especially in the presence of Gaussia-Dura luciferase reporter.


  • Highly sensitive detection of Gaussia luciferase activity
  • Increased signal stability
  • Assay reagents compatible with other Gaussia luciferases
  • Automation-friendly - amenable to high throughput assays
  • Contains a universal cell lysis buffer and optimized glow assay reagent
  • Allows one to perform non-radioactive assays


Light output generated by the luciferase reaction can be correlated to the amount of Gaussia luciferase protein produced and used to determine the activity of the promoter driving Gaussia expression


Kit may be stored at –80 ˚C, or individually at indicated temperatures.


The Gaussia Glow Assay Kit reagent yields a Gaussia-Dura luciferase reaction with glow-type kinetics and is recommended for luminometers without injectors or for applications requiring batch processing of samples.


The signal produced by Gaussia luciferase shows strong flash kinetics and is considerably greater than flash signals from either firefly or Renilla luciferases assayed under similar conditions.


Delivery information: Kit contains Gaussia glow assay buffer (5 or 50 ml) (store at 4 ˚C), 100X Coelenterazine (50 or 500 µl) (store at –80 ˚C) and 2X cell lysis buffer (6 or 60 ml) (store at room temperature).


Caution: For research use only. Not for use in diagnostic procedures.

Order Now

SPECIFICATIONS

Environmentally Preferable
Storage conditions –80 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR