Pierce™ Active GTPase Pull-down and Detection Kit, Thermo Scientific
Supplier: Thermo Scientific
16116 16117 16118 16119 16120 16121
CAPI16116EA 2512.21 CAD
CAPI16116 CAPI16117 CAPI16118 CAPI16119 CAPI16120 CAPI16121
Pierce™ Active GTPase Pull-down and Detection Kit, Thermo Scientific
Assays Enzyme Assays
The Thermo Scientific Active GTPase Pull-Down and Detection Kits allows the monitoring of changes in GTPase activity levels by enriching and detecting the active (GTP-bound) state of a particular GTPase.
  • Validated – functionally tested to ensure quality and performance
  • Sensitive – optimised antibodies, reagents and Western blotting procedure accurately detect changes in GTPase activity levels
  • Convenient – kits contain controls and all reagents needed to perform and detect 30 pull-downs
  • Easy to use – conditions are optimised for immediate success in a 2-hour assay
  • Efficient – spin columns prevent sample loss
These kits include all the necessary reagents to lyse cells, enrich for active GTPases, positive and negative controls and an anti-GTPase primary antibody for Western blot detection.
Delivery information: Each kit includes GST fusion protein of specific binding domain (1 vial), glutathione agarose resin (3 ml), GTPγS, 100X (50 µl), GDP 100X (50 µl), lysis/binding/wash buffer (100 ml), GTPase-specific primary antibody (1 vial), SDS-PAGE sample loading buffer, 2X (1,5 ml), spin columns (30), and collection tubes (90).
Caution: For Research Use Only. Not for use in diagnostic procedures.