Anti-MUC1 Rabbit Polyclonal Antibody

Supplier: Thermo Scientific
PA5-16847
CAPIPA5-16847EA 3242.11 CAD
CAPIPA5-16847
Anti-MUC1 Rabbit Polyclonal Antibody
Antibodies
This Antibody targets MUC-1 in IHC (P) applications and shows reactivity with Human and mouse samples. The immunogen is a synthetic peptide from cytoplasmic tail of human MUC-1 protein. MUC-1 is a heavily O-glycosylated transmembrane protein expressed on most secretory epithelium, including mammary gland and some hematopoietic cells. It is expressed abundantly in lactating mammary glands and overexpressed abundantly in >90% breast carcinomas and metastases. Transgenic MUC-1 has been shown to associate with all four cebB receptors and localize with erbB1 (EGFR) in lactating glands.

Recommended Dilutions: Immunohistochemistry (Paraffin): 1:50-100

Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR