Anti-EBF1/COE1 Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18720
CAPIPA5-18720EA 2904.29 CAD
CAPIPA5-18720
Anti-EBF1/COE1 Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with rat based on sequence homology. Transcriptional activator which recognizes variations of the palindromic sequence 5'-ATTCCCNNGGGAATT-3'.

Recommended Dilutions: Western Blot: 0.3-0.1 ug/ml, Immunoprecipitation: Assay Dependent

Type: Primary
Antigen: EBF1/COE1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR