Anti-RNLS Goat Polyclonal Antibody

Supplier: Thermo Scientific
PA5-18376
CAPIPA5-18376EA 3269.34 CAD
CAPIPA5-18376
Anti-RNLS Goat Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, canine and rat based on sequence homology. Renalase is a flavin adenine dinucleotide-dependent amine oxidase that is secreted into the blood from the kidney .

Recommended Dilutions: Western Blot: 0.1-0.3 ug/ml, Immunohistochemistry (Paraffin): 2-4 ug/ml

Type: Primary
Antigen: RNLS (renalase, FAD-dependent amine oxidase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR