Ethyl cyano(hydroxyimino)acetate
Supplier: BACHEM AMERICAS INC
4019721.0025
4019721.0100
Q-2750.0025BAEA
214.33
CAD
Q-2750.0025BA
Q-2750.0100BA
Ethyl cyano(hydroxyimino)acetate
Ethyl cyano(hydroxyimino)acetate
Highly efficient coupling additive, a safer, non-explosive alternative to HOBt.
Formula:
C₅H₆N₂O₃ |
MDL Number:
MFCD00002112 CAS Number: 3849-21-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...