Mca-Ala-Pro-Lys(Dnp)-Oh
Supplier: BACHEM AMERICAS INC
4042638.0001
4042638.0005
M-2710.0001BAEA
314.86
CAD
M-2710.0001BA
M-2710.0005BA
Mca-Ala-Pro-Lys(Dnp)-Oh
Proteins and Peptides
FRET substrate for angiotensin-converting enzyme 2 (ACE2). Mca-APK(Dnp)-OH is used for monitoring the activity of the enzyme in plasma, urine, heart, and lungs.