Ac-Lys-Gln-Leu-Arg-Afc I-2005.0005 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4049682.0001
4049682.0005
10796-732EA
183.83
CAD
10796-732
10796-734
Ac-Lys-Gln-Leu-Arg-Afc I-2005.0005 Trifluoroacetate
Proteins and Peptides
Hepsin, a membrane-anchored serine protease, is overexpressed in ovarian and prostate cancer and in renal cell carcinoma and thus can serve as prognostic marker. Ac-KQLR-AFC is the optimal substrate for human hepsin, as KQLR / VVNG corresponds to its cleavage site. Excitation at 395-400 nm, emission at 495-505 nm.