H-Pro-Pro-Asp-NH₂ Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4090413.0250 4090413.1000
10796-776EA 214.23 CAD
10796-776 10796-778
H-Pro-Pro-Asp-NH₂ Trifluoroacetate
Proteins and Peptides

Absorbed to modified silica gel, PPD-amide is a highly efficient recyclable chiral catalyst for aldol reactions.

Order Now

SPECIFICATIONS

Protein/peptide name H-Pro-Pro-Asp-NH2 Trifluoroacetate
Environmentally Preferable
Species
Source
CAS No. 850440-85-6
Sequence H-Pro-Pro-Asp-NH₂
Molecular weight 326.35
Molecular formula C₁₄H₂₂N₄O₅
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR