Lactoferricin B25 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4087382.0005 4087382.0001
10796-708EA 717.83 CAD
10796-708 10796-814
Lactoferricin B25 Trifluoroacetate
Proteins and Peptides
The antimicrobial peptide lactoferricin B25 is a fragment of cow's milk protein lactoferrin. The peptide exhibits potent dose-dependent and time-dependent anticancer activity against human stomach cancer cell cultures via apoptosis and autophagy. LFcinB25 is expected to yield novel anticancer drugs.
Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).