Fmoc-O-ethyl-L-tyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-ethoxyphenyl)propanoic acid, Fmoc-Tyr(4-Et)-OH

4043686.0001 4043686.0005
10796-858EA 355.07 CAD
10796-858 10796-860
Fmoc-O-ethyl-L-tyrosine
Fmoc-O-ethyl-L-tyrosine

Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine.Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₂₆H₂₅NO₅
MDL Number: MFCD02094093
CAS Number: 119894-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR