Fmoc-Dopa(acetonide)-OH
Supplier: BACHEM AMERICAS INC
4076682.0001
4076682.0005
10796-854EA
979.71
CAD
10796-854
10796-856
Fmoc-Dopa(acetonide)-OH
Fmoc-Dopa(acetonide)-OH
Protected derivative for the introduction of Dopa during Fmoc-SPPS. Dopa-containing peptides adhere to metal surfaces. The catechol moiety forms complexes with metal ions, e.g. with Fe (III).
Bachem offers derivatives of (di)halogenated tyrosines, (di)nitrotyrosine and other 3-substituted tyrosines.
Formula:
C₂₇H₂₅NO₆ |
MDL Number:
MFCD08064332 CAS Number: 852288-18-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...