Fmoc-Tyr(So2(Oneopentyl))-Oh
Supplier: BACHEM AMERICAS INC
Fmoc-Tyr(SO₃nP), a protected derivative of sulfotyrosine. The neopentyl ester is stable under the conditions of Fmoc-SPPS including the TFA-cleavage, but can be removed by treatment with aqueous ammonium acetate or sodium acetate. Thus, premature loss of sulfate groups during the final deprotection is prevented.
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine.Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.