Fmoc-Cys(Dpm)-OH

Supplier: BACHEM AMERICAS INC

4081840.0005 4081840.0025
B-4225.0005BAEA 256.51 CAD
B-4225.0005BA B-4225.0025BA
Fmoc-Cys(Dpm)-OH
Fmoc-Cys(Dpm)-OH

An alternative to Mob in the regioselective formation of disulfide bonds.

Formula: C₃₁H₂₇NO₄S
MDL Number: MFCD02682541
CAS Number: 247595-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR