Fmoc-N-Me-Ile-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-isoleucine, Fmoc-N-Me-Ile-OH, N-Fmoc-N-methyl-L-isoleucine
4042030.0001
4042030.0005
B-4215.0001BAEA
196.13
CAD
B-4215.0001BA
B-4215.0005BA
Fmoc-N-Me-Ile-OH
Fmoc-N-Me-Ile-OH
MeIle derivative used in the Fmoc-SPPS of cyclosporin and compstatin analogs.
Formula:
C₂₂H₂₅NO₄ MW: 367.445 g/mol |
MDL Number:
MFCD00153389 CAS Number: 138775-22-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...