Fmoc-L-Phe(4-Br)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Phe(4-Br)-OH

4042637.0001 4042637.0005
B-4200.0001BAEA 250.37 CAD
B-4200.0001BA B-4200.0005BA
Fmoc-L-Phe(4-Br)-OH
Fmoc-L-Phe(4-Br)-OH

Educt (coupled to Wang resin) for obtaining a library of p-biphenylalanines via Suzuki coupling.


Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.

Formula: C₂₄H₂₀BrNO₄
MW: 466.30 g/mol
MDL Number: MFCD00273460
CAS Number: 198561-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR