Fmoc-L-allylglycine
Supplier: BACHEM AMERICAS INC
4074454.0001
4074454.0025
B-4190.0001BAEA
377.3
CAD
B-4190.0001BA
B-4190.0025BA
Fmoc-L-allylglycine
Fmoc-L-allylglycine
Allylglycine-containing peptides may be cleaved selectively at the amide bond between allylglycine and the subsequent amino acid with iodine. The lateral double bond allows selective modifications of a peptide e.g. via metathesis. Replacing cysteine residues by allylglycine allows to obtain carba-analogs of disulfide-bridged peptides.Educt for obtaining Fmoc-prenylglycine.
Formula:
C₂₀H₁₉NO₄ |
MDL Number:
MFCD01311749 CAS Number: 146549-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...