Fmoc-(Dmb)Ala-OH
Supplier: BACHEM AMERICAS INC
4072316.1000
4072316.5000
B-4175.1000BAEA
627.31
CAD
B-4175.1000BA
B-4175.5000BA
Fmoc-(Dmb)Ala-OH
Fmoc-(Dmb)Ala-OH
Derivative for the Fmoc-SPPS of backbone-protected peptides. Introduction of Dmb at appropriate positions prevents aggregation of the growing peptide chain. Conroy et al. could stabilize the aspartimide-prone Asp(OAll or ODmab)-Ala motif by incorporating DmbAla. Please see also our Dmb-dipeptides.
Bachem additionally offers deuterated alanine: Fmoc-[D₄]Ala-OH (B-4130).
Formula:
C₂₇H₂₇NO₆ Storage Temperature: Ambient |
MDL Number:
MFCD18910716 CAS Number: 1425938-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...