4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
4-Azido-N-Fmoc-L-homoalanine
4072396.0250
4072396.1000
B-4145.0250BAEA
500.88
CAD
B-4145.0250BA
B-4145.1000BA
4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine
4-Azido-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-homoalanine
Protected derivative of L-azidohomoalanine (Aha), a Met isostere. This building block for 'click chemistry' additionally creates a specific cleavage site in peptides, Aha-containing sequences can be cleaved under mild conditions in the presence of phosphines or dithiols via formation of homoserine lactone.
Formula:
C₁₉H₁₈N₄O₄ MW: 366.38 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD11052920 CAS Number: 942518-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...