Fmoc-Ala-OH (2,3,3,3-D4)

Supplier: BACHEM AMERICAS INC

4071077.0500 4071077.1000
B-4130.0500BAEA 627.31 CAD
B-4130.0500BA B-4130.1000BA
Fmoc-Ala-OH (2,3,3,3-D4)
Fmoc-Ala-OH (2,3,3,3-D4)

Fmoc-(D₄)-Ala-OH was used by Bowman and Zaia in the SPPS of peptide tags for the stable isotopic labeling of carbohydrates which allowed their mass spectroscopic quantification. For ²H-NMR-studies on the influence of Pro on the folding of WALP transmembrane peptides, Koeppe’s group incorporated pairs of deuterated alanines by introducing Fmoc-(D₄)-Ala-OH (60-100%) at selected sequence positions.

Formula: C₁₈H₁₃D₄NO₄
MW: 315.31 g/mol
MDL Number: MFCD03428190
CAS Number: 225101-69-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR