Fmoc-4-Sulfomethyl-Phe(Tce)-Oh
Supplier: BACHEM AMERICAS INC
4070723.0001 4070723.0005
B-4110.0001BAEA 1505.11 CAD
B-4110.0001BA B-4110.0005BA
Fmoc-4-Sulfomethyl-Phe(Tce)-Oh
Proteins and Peptides
Derivative of a stable sulfotyrosine mimic suitable for Fmoc-SPPS. The lateral trichloroethyl (Tce) group is cleaved by hydrogenolysis (H₂/Pd in the presence of ammonium formate).
Bachem offers a considerable choice of ring-substituted phenylalanines, protected derivatives of: p-Aminophenylalanine p-Azidophenylalanine p-Benzoylphenylalanine p-Biphenylalanine p-Bromophenylalanine p-t-Butylphenylalanine p-Carboxyphenylalanine p-Chlorophenylalanine p-Cyanophenylalanine 3,4-Dichlorophenylalanine Fluorophenylalanines p-Iodophenylalanine p-Methylphenylalanine p-Nitrophenylalanine p-Phosphonophenylalanine. p-Methoxy- and p-ethoxyphenylalanine derivatives are listed in the Tyr family, whereas β,β-diphenylalanine derivatives can be found in the "β-Substituted Alanines" family.