Fmoc-Arg(Me,Pbf)-OH

Supplier: BACHEM AMERICAS INC

4043408.0500 4043408.1000
B-4010.0500BAEA 1378.68 CAD
B-4010.0500BA B-4010.1000BA
Fmoc-Arg(Me,Pbf)-OH
Fmoc-Arg(Me,Pbf)-OH

Nω-Methylation is a widespread post-translational modification of Arg-containing proteins. Arg(Me) can be conveniently introduced during the Fmoc-SPPS of model peptides using this derivative.


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₅H₄₂N₄O₇S
MDL Number: MFCD08064312
CAS Number: 1135616-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR