Pg 97-269 Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4048647.0500 4048647.1000
H-7286.0500BAEA 650.47 CAD
H-7286.0500BA H-7286.1000BA
Pg 97-269 Trifluoroacetate
Proteins and Peptides
PG 97-269 is a selective high affinity VPAC1 receptor antagonist with negligible affinity for the PACAP type I receptor. The Ki values were 15 +/- 5 nM for the rat and 2 +/- 1 nM for the human VPAC1 receptors. PG 97-269 may be useful for the evaluation of the physiological role of VIP in rat and human tissues.