Human ((13C6)Leu14)-Glucagon (1-29) Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4072022.0500 4072022.1000
H-7236.0500BAEA 627.31 CAD
H-7236.0500BA H-7236.1000BA
Human ((13C6)Leu14)-Glucagon (1-29) Trifluoroacetate
Proteins and Peptides
Heavy isotope-labeled Glucagon useful for pharmacokinetic/pharmacodynamic studies in combination with Bachem product H-6790 Glucagon (1-29) (human, rat, porcine) or the corresponding generic API (Glucagon).
For the long-acting GLP-1 analog liraglutide see H-6724.