Cyclo(-Arg-Gly-Asp-D-Phe-Cys) Acetate
Supplier: BACHEM AMERICAS INC
4069272.0001
4069272.0005
H-7226.0001BAEA
211.72
CAD
H-7226.0001BA
H-7226.0005BA
Cyclo(-Arg-Gly-Asp-D-Phe-Cys) Acetate
Proteins and Peptides
c(RGDfC), a RGD tumor-targeting peptide, which binds to αvβ3 receptors. c(RGDfC) can be easily conjugated to drugs as combretastatin or surfaces. For the control peptide c(RADfC) see H-7232