Fmoc-Cys(Pam)2-OH(S)
Supplier: BACHEM AMERICAS INC
4074791.0500
4074791.1000
10796-862EA
1712.77
CAD
10796-862
10796-864
Fmoc-Cys(Pam)2-OH(S)
Fmoc-Cys(Pam)2-OH(S)
SPPS employing the pure stereoisomer of Fmoc-Pam₂Cys-OH allows to obtain more homogeneous lipopeptides. The configuration of the bis-palmitoyloxypropyl moiety could influence the biological activity of the peptide conjugate.
Formula:
C₅₃H₈₃NO₈S |
MDL Number:
MFCD30742871 CAS Number: 139573-78-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...