Fmoc-Cys(Pam)2-OH(R)

Supplier: BACHEM AMERICAS INC

4042951.0500 4042951.1000
10796-866EA 1836.09 CAD
10796-866 10796-868
Fmoc-Cys(Pam)2-OH(R)
Fmoc-Cys(Pam)2-OH(R)

SPPS employing the pure stereoisomer of Fmoc-Pam₂Cys-OH allows to obtain more homogeneous lipopeptides. The configuration of the bis-palmitoyloxypropyl moiety could influence the biological activity of the peptide conjugate. For Fmoc-(S)-Pam₂Cys-OH see B-4335.

Formula: C₅₃H₈₃NO₈S
MW: 894.3 g/mol
CAS Number: 139573-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR