Fmoc-Oic-OH

Supplier: BACHEM AMERICAS INC

4025301.0250 4025301.1000
B-2425.0250BAEA 180.83 CAD
B-2425.0250BA B-2425.1000BA B-2425.5000BA
Fmoc-Oic-OH
Fmoc-Oic-OH
Formula: C₂₄H₂₅NO₄
MDL Number: MFCD00191201
CAS Number: 130309-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR