H-G-Azido-Abu-Oh
Supplier: BACHEM AMERICAS INC
4075013.0250
4075013.1000
F-4265.0250BAEA
475.07
CAD
F-4265.0250BA
F-4265.1000BA
H-G-Azido-Abu-Oh
Proteins and Peptides
The methionine isostere γ-azidohomoalanine is fed to bacteria for obtaining “clickable” proteins by recombinant synthesis. Additionally, the azido moiety can serve as a probe for studying protein conformations by IR-spectroscopy.