Boc-Lys(N3)-OH DCHA
Supplier: BACHEM AMERICAS INC
4072397.0250
4072397.1000
A-4765.0250BAEA
377.3
CAD
A-4765.0250BA
A-4765.1000BA
Boc-Lys(N3)-OH DCHA
Boc-Lys(N3)-OH DCHA
Lysine-derived building block for "click chemistry" and other reactions involving azides.The azido moiety may also be selectively converted into an amino group with triphenylphosphine in the presence of water.
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₁₁H₂₀N₄O₄·C₁₂H₂₃N MW: 453.63 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD12546084 CAS Number: 846549-33-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...