Boc-L-octahydroindole-2-carboxylic acid

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Oic-OH, (2S,3aS,7aS)-1-(tert-Butoxycarbonyl)octahydro-1H-indole-2-carboxylic acid, Boc-(2S,3aS,7aS)-octahydro-1H-indole-2-carboxylic acid

4025299.1000 4025299.5000
A-3645.0250BAEA 161.78 CAD
A-3645.0250BA A-3645.1000BA A-3645.5000BA
Boc-L-octahydroindole-2-carboxylic acid
Boc-L-octahydroindole-2-carboxylic acid
Formula: C₁₄H₂₃NO₄
MDL Number: MFCD09750486
CAS Number: 109523-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR