Anti-TACC2 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-5736R
10422-324EA 1057.32 CAD
10422-324
Anti-TACC2 Rabbit Polyclonal Antibody
Antibodies
TACC2 is a conserved family of centrosome and microtubule interacting proteins that are implicated in cancer. This gene encodes a protein that concentrates at centrosomes throughout the cell cycle. This gene lies within a chromosomal region associated with tumorigenesis. Expression of this gene is thought to affect the progression of breast tumors. Expression of this gene is also induced by erythropoietin.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TACC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR