Anti-GTF2B Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-8168R-CY5
10478-364EA 1288.98 CAD
10478-364
Anti-GTF2B Rabbit Polyclonal Antibody (Cy5®)
Antibodies
General factor that plays a major role in the activation of eukaryotic genes transcribed by RNA polymerase II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GTF2B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR