Anti-F10 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-7762R-CY5
10468-920EA 1288.62 CAD
10468-920
Anti-F10 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Factor Xa is a vitamin K-dependent glycoprotein that converts prothrombin to thrombin in the presence of factor Va, calcium and phospholipid during blood clotting.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: F10
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR