Anti-Ribonuclease Inhibitor Rabbit Polyclonal Antibody

Supplier: Bioss

BS-7518R
10463-768EA 1000.39 CAD
10463-768
Anti-Ribonuclease Inhibitor Rabbit Polyclonal Antibody
Antibodies
Ribonuclease inhibitor which inhibits RNASE1, RNASE2 and ANG. May play role in redox homeostasis.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: Ribonuclease Inhibitor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR