Anti-NUPR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-7106R-A555
10461-866EA 1288.98 CAD
10461-866
Anti-NUPR1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Could participate in the response to proapoptotic stimuli and promotes cellular growth in a way that helps the tissue counteract diverse injuries. May contribute to the metastatic phenotype.Tissue specificity: Highly expressed in pancreas.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NUPR1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR