Anti-MAGEA5 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-6820R-CY3
10455-392EA 1288.98 CAD
10455-392
Anti-MAGEA5 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
MAGEA5 is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. This MAGEA gene encodes a protein that is C-terminally truncated compared to other family members, and this gene can be alternatively interpreted to be a pseudogene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAGEA5
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR