Anti-PP2A beta Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-6706R-CY3
10452-966EA 1288.98 CAD
10452-966
Anti-PP2A beta Rabbit Polyclonal Antibody (Cy3®)
Antibodies
PP2A can modulate the activity of phosphorylase B kinase casein kinase 2, mitogen-stimulated S6 kinase, and MAP-2 kinase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PP2A beta
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR