Anti-FGFR1OP Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-5712R-CY5
10421-818EA 1320 CAD
10421-818
Anti-FGFR1OP Rabbit Polyclonal Antibody (Cy5®)
Antibodies
FGFR1 Oncogene Partner is required for anchoring microtubules to the centrosomes. Ubiquitous; highly expressed in heart, liver, muscle, kidney, intestine, colon, adrenal gland, prostate, testis, and pancreas. A chromosomal aberration involving FGFR1OP may be a cause of stem cell myeloproliferative disorder (MPD). There are three named isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGFR1OP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR