Anti-AGPAT4 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-5033R-CY5
10406-456EA 1320 CAD
10406-456
Anti-AGPAT4 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Converts lysophosphatidic acid (LPA) into phosphatidic acid by incorporating an acyl moiety at the sn-2 position of the glycerol backbone.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AGPAT4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR