Anti-CD5L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-2487R-FITC
10339-852EA 1320 CAD
10339-852
Anti-CD5L Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
May play a role in the regulation of the immune system. Seems to play a role as an inhibitor of apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD5L
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR